Product Details

SNP ID
rs9757
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:137875951 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTTTTCCTGTGTTAAGTGCTGACC[C/G]TTTGACCACAAAGGCATGGAACATT
Phenotype
MIM: 608834
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CREB3L2 PubMed Links

Gene Details

Gene
CREB3L2
Gene Name
cAMP responsive element binding protein 3 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253775.1 6071 Intron NP_001240704.1
NM_001318246.1 6071 UTR 3 NP_001305175.1
NM_194071.3 6071 UTR 3 NP_919047.2
XM_005250541.4 6071 UTR 3 XP_005250598.1
XM_017012540.1 6071 UTR 3 XP_016868029.1
XM_017012541.1 6071 UTR 3 XP_016868030.1

View Full Product Details