Product Details

SNP ID
rs11725902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:75576770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCAGCCTTATATTGTAGCACAA[A/C]TAAGATAAAGGAACCAATAATGAGA
Phenotype
MIM: 603442
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDKL2 PubMed Links

Gene Details

Gene
CDKL2
Gene Name
cyclin dependent kinase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263335.2 3745 UTR 3 XP_005263392.1
XM_006714406.2 3745 UTR 3 XP_006714469.1
XM_017008809.1 3745 UTR 3 XP_016864298.1
XM_017008810.1 3745 UTR 3 XP_016864299.1
XM_017008811.1 3745 UTR 3 XP_016864300.1

View Full Product Details