Product Details
- SNP ID
-
rs7017914
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:70678968 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCATAGCAGAATACCACAGACTT[A/G]TTAGCTTAAACAATAGAAATTTATT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LACTB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143606190] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LACTB2
- Gene Name
- lactamase beta 2
There are no transcripts associated with this gene.
- Gene
- XKR9
- Gene Name
- XK related 9
View Full Product Details