Product Details

SNP ID
rs2618481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11496588 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCTCTGTGGCAGGGGCTGCAGGA[C/T]GGAATACACTCTCATCTGCCCAGCG
Phenotype
MIM: 191305
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BLK PubMed Links

Gene Details

Gene
BLK
Gene Name
BLK proto-oncogene, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001715.2 Intron NP_001706.2
XM_011543824.1 Intron XP_011542126.1
XM_011543825.2 Intron XP_011542127.1
XM_011543827.1 Intron XP_011542129.1
XM_011543828.2 Intron XP_011542130.1
XM_011543829.2 Intron XP_011542131.1
XM_017013730.1 Intron XP_016869219.1

View Full Product Details