Product Details

SNP ID
rs2025348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:131127624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCACATTATAAACTTCTATATAC[G/T]AATTAAAATACAAATAACAAACTAG
Phenotype
MIM: 604693
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AKAP7 PubMed Links

Gene Details

Gene
AKAP7
Gene Name
A-kinase anchoring protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004842.3 Intron NP_004833.1
NM_016377.3 Intron NP_057461.2
NM_138633.2 Intron NP_619539.1
XM_005267228.1 Intron XP_005267285.1
XM_005267229.4 Intron XP_005267286.1
XM_006715614.2 Intron XP_006715677.1
XM_011536262.1 Intron XP_011534564.1
XM_017011506.1 Intron XP_016866995.1
XM_017011507.1 Intron XP_016866996.1
XM_017011508.1 Intron XP_016866997.1
XM_017011509.1 Intron XP_016866998.1
XM_017011510.1 Intron XP_016866999.1
XM_017011511.1 Intron XP_016867000.1

View Full Product Details