Product Details

SNP ID
rs10787760
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:117131182 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGAGAGAGAGACCTGGAAGGAAGAA[A/G]TAATGGAGGAGGGAGACAGGGGGAA
Phenotype
MIM: 611171 MIM: 604294
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHTN1 PubMed Links

Gene Details

Gene
SHTN1
Gene Name
shootin 1
There are no transcripts associated with this gene.

Gene
VAX1
Gene Name
ventral anterior homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112704.1 1833 Intron NP_001106175.1
NM_199131.2 1833 UTR 3 NP_954582.1

View Full Product Details