Product Details

SNP ID
rs6473939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:54511372 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTAGAGTTTTTTACATGTGTATCT[A/C]ATTTTTTAAGTGGCTGCATAGTATC
Phenotype
MIM: 603937
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RP1 PubMed Links

Gene Details

Gene
RP1
Gene Name
retinitis pigmentosa 1 (autosomal dominant)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006269.1 Intron NP_006260.1
XM_017013721.1 Intron XP_016869210.1
XM_017013722.1 Intron XP_016869211.1

View Full Product Details