Product Details
- SNP ID
-
rs11838784
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:51335201 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGATTTCTCTTCAACAAAAGACAT[A/C]ATGAAGAGAGTAAATAGATAATGGC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SERPINE3
PubMed Links
Gene Details
- Gene
- SERPINE3
- Gene Name
- serpin family E member 3
View Full Product Details