Product Details

SNP ID
rs10489770
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:46341925 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTAGCGGATGGCCTTCAGCTTTT[A/G]GAACCAGCCCGGCAACCTCCTTTCC
Phenotype
MIM: 615394
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSUN4 PubMed Links

Gene Details

Gene
NSUN4
Gene Name
NOP2/Sun RNA methyltransferase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256127.1 Intron NP_001243056.1
NM_001256128.1 Intron NP_001243057.1
NM_199044.3 Intron NP_950245.2

View Full Product Details