Product Details

SNP ID
rs518686
Assay Type
Functionally tested
NCBI dbSNP Submissions
77
Location
Chr.1:231241335 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GCCCCCAGCAGGAGCACCACCACGG[C/T]TTAGCAAAGAATCCCAGACCCCGCC
Phenotype
MIM: 602744
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf131 PubMed Links

Gene Details

Gene
C1orf131
Gene Name
chromosome 1 open reading frame 131
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300830.1 163 Intron NP_001287759.1
NM_152379.3 163 Intron NP_689592.2
Gene
GNPAT
Gene Name
glyceronephosphate O-acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316350.1 163 UTR 5 NP_001303279.1
NM_014236.3 163 UTR 5 NP_055051.1
XM_005273313.4 163 UTR 5 XP_005273370.1
XM_011544303.2 163 UTR 5 XP_011542605.1
XM_011544304.2 163 UTR 5 XP_011542606.1

View Full Product Details