Product Details

SNP ID
rs35808381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53963671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGCAGCCCCTCTGCCTCTCCG[A/C]TGAACCCCCTTATCCTATCTTTCCC
Phenotype
MIM: 606900
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CACNG8 PubMed Links

Gene Details

Gene
CACNG8
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031895.5 Intron NP_114101.4

View Full Product Details