Product Details

SNP ID
rs735811
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:8369893 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACTCACAGCCAGGGGACCTCAGGCA[C/T]GGGACATGAGCATTGACACGCCGAG
Phenotype
MIM: 603402
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOX3 PubMed Links

Gene Details

Gene
ACOX3
Gene Name
acyl-CoA oxidase 3, pristanoyl
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101667.1 Intron NP_001095137.1
NM_003501.2 Intron NP_003492.2
XM_005248011.4 Intron XP_005248068.1
XM_005248013.3 Intron XP_005248070.1
XM_011513565.2 Intron XP_011511867.1
XM_011513566.1 Intron XP_011511868.1

View Full Product Details