Product Details
- SNP ID
-
rs35381145
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:3076567 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCAACACCCCCCACCCCGCTTGT[G/T]ATTTTTCTCCTTAGCACTTACGGCC
- Phenotype
-
MIM: 171840
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101927824
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75981723] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101927824
- Gene Name
- translation initiation factor IF-2-like
There are no transcripts associated with this gene.
- Gene
- PFKP
- Gene Name
- phosphofructokinase, platelet
View Full Product Details