Product Details

SNP ID
rs11965090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:17763113 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTTCAGAGCAGAATGCTCAGAAG[C/G]AGAACATCTCTTATTTATGTAATTT
Phenotype
MIM: 605433
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIF13A PubMed Links

Gene Details

Gene
KIF13A
Gene Name
kinesin family member 13A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105566.2 Intron NP_001099036.1
NM_001105567.2 Intron NP_001099037.1
NM_001105568.2 Intron NP_001099038.1
NM_001243423.1 Intron NP_001230352.1
NM_022113.5 Intron NP_071396.4
XM_011514806.2 Intron XP_011513108.1
XM_017011188.1 Intron XP_016866677.1
XM_017011189.1 Intron XP_016866678.1
XM_017011190.1 Intron XP_016866679.1
XM_017011191.1 Intron XP_016866680.1
XM_017011192.1 Intron XP_016866681.1
XM_017011193.1 Intron XP_016866682.1

View Full Product Details