Product Details

SNP ID
rs12872770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:108497555 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACCCAGAGTGTGATATTTATTTA[C/T]TGAAGGAAAAATGTATTTAACCAAT
Phenotype
MIM: 615479
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO16 PubMed Links

Gene Details

Gene
MYO16
Gene Name
myosin XVI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198950.1 Intron NP_001185879.1
NM_015011.1 Intron NP_055826.1
XM_011521062.1 Intron XP_011519364.1

View Full Product Details