Product Details

SNP ID
rs3746721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1630248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCAGGTCTTCTGCTTCCAGGGGAC[A/G]TAGATGGGGCCCAGGAGGACAGCTA
Phenotype
MIM: 603889 MIM: 605466
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIRPB1 PubMed Links

Gene Details

Gene
SIRPB1
Gene Name
signal regulatory protein beta 1
There are no transcripts associated with this gene.

Gene
SIRPG
Gene Name
signal regulatory protein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039508.1 872 Silent Mutation TAC,TAT Y269Y NP_001034597.1
NM_018556.3 872 Silent Mutation TAC,TAT Y380Y NP_061026.2
NM_080816.2 872 Silent Mutation TAC,TAT Y163Y NP_543006.2
XM_005260749.3 872 Intron XP_005260806.1
XM_011529286.2 872 Silent Mutation TAC,TAT Y347Y XP_011527588.1
XM_011529287.2 872 Intron XP_011527589.1
Gene
SIRPG-AS1
Gene Name
SIRPG antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details