Product Details

SNP ID
rs11790228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136113043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGTTTGGTGAAACAGTTCCACA[A/T]AACTCTACAATGGATAGGTTGTTAC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C9orf69 PubMed Links

Gene Details

Gene
C9orf69
Gene Name
chromosome 9 open reading frame 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256526.1 Intron NP_001243455.1
NM_152833.2 Intron NP_690046.3
Gene
LOC105376323
Gene Name
uncharacterized LOC105376323
There are no transcripts associated with this gene.

Gene
LOC107987142
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

View Full Product Details