Product Details

SNP ID
rs4032696
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:73423989 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGTGCAAAATATATGAACAGGAAA[A/T]ATTTAACAAACAGCAGTTGCCTCTA
Phenotype
MIM: 606286 MIM: 613973
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DDX43 PubMed Links

Gene Details

Gene
DDX43
Gene Name
DEAD-box helicase 43
There are no transcripts associated with this gene.

Gene
MB21D1
Gene Name
Mab-21 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138441.2 1923 Intron NP_612450.2
XM_017010232.1 1923 UTR 3 XP_016865721.1

View Full Product Details