Product Details

SNP ID
rs1408479
Assay Type
Validated
NCBI dbSNP Submissions
53
Location
Chr.1:59894355 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGGCATGGGTGGCCCTCTGTATTG[C/T]GACTGCTTTGGCCTTCCTCCTTCAG
Phenotype
MIM: 601258
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP2J2 PubMed Links

Gene Details

Gene
CYP2J2
Gene Name
cytochrome P450 family 2 subfamily J member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000775.3 Intron NP_000766.2

View Full Product Details