Product Details

SNP ID
rs535135
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:64719383 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTCAAGCAAAAGATAAATGACAAG[C/G]TTCCTGCCACCAGCAGCTCAAATGC
Phenotype
MIM: 607664
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GNS PubMed Links

Gene Details

Gene
GNS
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002076.3 Intron NP_002067.1

View Full Product Details