Product Details

SNP ID
rs4713658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33699340 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCGGACCAATGGCGGGACTATCTC[C/G]CAGCCTTCGTTGCTGCCTGTTTTCC
Phenotype
MIM: 147267 MIM: 614461
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITPR3 PubMed Links

Gene Details

Gene
ITPR3
Gene Name
inositol 1,4,5-trisphosphate receptor type 3
There are no transcripts associated with this gene.

Gene
MIR3934
Gene Name
microRNA 3934
There are no transcripts associated with this gene.

Gene
UQCC2
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032340.3 Intron NP_115716.1

View Full Product Details