Product Details

SNP ID
rs7324458
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:52134637 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCTTCTAAAAGGCATTTCAAAATAG[A/C]ACTATGACATATTTTTTAAACAGGG
Phenotype
MIM: 604044 MIM: 616731
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929657 PubMed Links

Gene Details

Gene
LOC101929657
Gene Name
uncharacterized LOC101929657
There are no transcripts associated with this gene.

Gene
NEK3
Gene Name
NIMA related kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146099.1 Intron NP_001139571.1
NM_002498.2 Intron NP_002489.1
NM_152720.2 Intron NP_689933.1
Gene
NEK5
Gene Name
NIMA related kinase 5
There are no transcripts associated with this gene.

View Full Product Details