Product Details

SNP ID
rs11580979
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:157999516 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGCAAAGGGGTGGCAGTTGGTAT[G/T]GAGGGAAGGGATGGGGACAAATTCC
Phenotype
MIM: 607428
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIRREL PubMed Links

Gene Details

Gene
KIRREL
Gene Name
kin of IRRE like (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286349.1 Intron NP_001273278.1
NM_018240.6 Intron NP_060710.3
XM_005245305.4 Intron XP_005245362.1

View Full Product Details