Product Details
- SNP ID
-
rs12459144
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:10318114 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACTCCACCCTGAGCAAGACCAACC[C/T]CTGCCACCACCCCAGCTTTGGCCTG
- Phenotype
-
MIM: 614585
MIM: 609950
MIM: 611639
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FDX1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77438995] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FDX1L
- Gene Name
- ferredoxin 1 like
There are no transcripts associated with this gene.
- Gene
- RAVER1
- Gene Name
- ribonucleoprotein, PTB binding 1
- Gene
- ZGLP1
- Gene Name
- zinc finger, GATA-like protein 1
There are no transcripts associated with this gene.
View Full Product Details