Product Details

SNP ID
rs11866378
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:70292564 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATATTTTTCTCTTGTAGATGTCAAC[A/T]ACATGCTAGAAAGAATGCTATCTTA
Phenotype
MIM: 601065 MIM: 605812
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AARS PubMed Links

Gene Details

Gene
AARS
Gene Name
alanyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
DDX19B
Gene Name
DEAD-box helicase 19B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014449.2 Intron NP_001014449.1
NM_001014451.2 Intron NP_001014451.1
NM_001257172.1 Intron NP_001244101.1
NM_001257173.1 Intron NP_001244102.1
NM_001257174.1 Intron NP_001244103.1
NM_001257175.1 Intron NP_001244104.1
NM_007242.5 Intron NP_009173.1
XM_006721127.2 Intron XP_006721190.1
XM_011522832.2 Intron XP_011521134.1
XM_011522833.2 Intron XP_011521135.1
XM_011522834.2 Intron XP_011521136.1
XM_017022890.1 Intron XP_016878379.1

View Full Product Details