Product Details

SNP ID
rs2634682
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:48339160 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATATTGGTGACTAGCCCCAGTTCTT[C/T]TACTGAGTTAAGAGTGGAAGAAGAC
Phenotype
MIM: 613906
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
H1FNT PubMed Links

Gene Details

Gene
H1FNT
Gene Name
H1 histone family member N, testis specific
There are no transcripts associated with this gene.

Gene
ZNF641
Gene Name
zinc finger protein 641
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172681.1 5063 Intron NP_001166152.1
NM_001172682.1 5063 Intron NP_001166153.1
NM_152320.2 5063 Intron NP_689533.2
XM_005268637.3 5063 UTR 3 XP_005268694.1
XM_005268638.3 5063 UTR 3 XP_005268695.1
XM_005268639.2 5063 UTR 3 XP_005268696.1
XM_005268640.4 5063 Intron XP_005268697.1
XM_011537897.1 5063 UTR 3 XP_011536199.1
XM_011537898.1 5063 UTR 3 XP_011536200.1
XM_017018797.1 5063 UTR 3 XP_016874286.1
XM_017018798.1 5063 UTR 3 XP_016874287.1
XM_017018799.1 5063 Intron XP_016874288.1

View Full Product Details