Product Details
- SNP ID
-
rs1799403
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:126440527 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACCCCCTACTTGCTCCTGTTCTGCC[A/G]GTGGCACCAGCTTCCCTACCGTCTC
- Phenotype
-
MIM: 615746
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CFAP100
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116252757] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CFAP100
- Gene Name
- cilia and flagella associated protein 100
There are no transcripts associated with this gene.
- Gene
- LOC105374089
- Gene Name
- uncharacterized LOC105374089
There are no transcripts associated with this gene.
- Gene
- ZXDC
- Gene Name
- ZXD family zinc finger C
View Full Product Details