Product Details

SNP ID
rs9857786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126609902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACAATGTGTACAGTCTCTCCCT[A/G]CTCCCTTTGGAAAGGCCGCCCACAA
Phenotype
MIM: 606235
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TXNRD3 PubMed Links

Gene Details

Gene
TXNRD3
Gene Name
thioredoxin reductase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173513.1 Intron NP_001166984.1
NM_052883.1 Intron NP_443115.1
Gene
TXNRD3NB
Gene Name
thioredoxin reductase 3 neighbor
There are no transcripts associated with this gene.

View Full Product Details