Product Details
- SNP ID
-
rs1235
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:46840293 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TATATTCCAATTGCTTGCATAATCA[G/T]TTTTTTTAATCCTGGGGTGTTGAAA
- Phenotype
-
MIM: 611128
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MDGA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs57253420] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MDGA2
- Gene Name
- MAM domain containing glycosylphosphatidylinositol anchor 2
View Full Product Details