Product Details
- SNP ID
-
rs1881270
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:62328943 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTCAGGCTAAGGAATTCACACTTC[A/G]TCCTCTATGTAGTGGGTACTGATGA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C3orf14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs538365662] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C3orf14
- Gene Name
- chromosome 3 open reading frame 14
- Gene
- PTPRG-AS1
- Gene Name
- PTPRG antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details