Product Details

SNP ID
rs1681248
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:98624535 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGGCGTCCTTGGGGCCCTGTACTT[C/G]CTGTCACCTGTGTCTGGGGTTCTCT
Phenotype
MIM: 600750
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NPTX2 PubMed Links

Gene Details

Gene
NPTX2
Gene Name
neuronal pentraxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002523.2 Intron NP_002514.1

View Full Product Details