Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001136000.2 | 7302 | UTR 3 | NP_001129472.1 | ||
NM_001136001.1 | 7302 | Intron | NP_001129473.1 | ||
NM_001168236.1 | 7302 | UTR 3 | NP_001161708.1 | ||
NM_001168237.1 | 7302 | UTR 3 | NP_001161709.1 | ||
NM_001168238.1 | 7302 | UTR 3 | NP_001161710.1 | ||
NM_001168239.1 | 7302 | UTR 3 | NP_001161711.1 | ||
NM_005158.4 | 7302 | UTR 3 | NP_005149.4 | ||
NM_007314.3 | 7302 | UTR 3 | NP_009298.1 | ||
XM_005245088.2 | 7302 | Intron | XP_005245145.1 | ||
XM_017001035.1 | 7302 | Intron | XP_016856524.1 |