Product Details
- SNP ID
-
rs610149
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
26
- Location
-
Chr.1:84929933 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAGAGCCAGGGAGAGGTTTTTTT[A/T]AAAAAAAAAAAAGGTAATGAAGGAA
- Phenotype
-
MIM: 607399
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MCOLN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs550754228] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MCOLN2
- Gene Name
- mucolipin 2
View Full Product Details