Product Details

SNP ID
rs12068735
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:111230651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTCTGGCAGAATGAGCAAATGA[C/T]GCAATGGCTGTCTTGGGGAGAAAAT
Phenotype
MIM: 601526
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHI3L2 PubMed Links

Gene Details

Gene
CHI3L2
Gene Name
chitinase 3 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025197.1 Intron NP_001020368.1
NM_001025199.1 Intron NP_001020370.1
NM_004000.2 Intron NP_003991.2

View Full Product Details