Product Details

SNP ID
rs4796583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42403241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCACCATGCTCCTAGGGAAGGAG[C/G]TTGAGAAGAAACTGCCAGGAGTGAA
Phenotype
MIM: 603198
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTRF PubMed Links

Gene Details

Gene
PTRF
Gene Name
polymerase I and transcript release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012232.5 2842 UTR 3 NP_036364.2
XM_005257242.3 2842 Intron XP_005257299.1

View Full Product Details