Product Details

SNP ID
rs1905455
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:58355463 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAATTGATATTTGAGGTAACTGTGG[G/T]AAGCAACGTAATGGGTTTTGAGGAA
Phenotype
MIM: 602961
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2D1 PubMed Links

Gene Details

Gene
UBE2D1
Gene Name
ubiquitin conjugating enzyme E2 D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204880.1 Intron NP_001191809.1
NM_003338.4 Intron NP_003329.1
XM_017016609.1 Intron XP_016872098.1

View Full Product Details