Product Details

SNP ID
rs11712445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47393062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGCCCAGTCTGGCCTTGAACTC[C/T]TGGGGTCAAGTGATCCTCTGGCCTC
Phenotype
MIM: 606584
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPN23 PubMed Links

Gene Details

Gene
PTPN23
Gene Name
protein tyrosine phosphatase, non-receptor type 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304482.1 Intron NP_001291411.1
NM_015466.3 Intron NP_056281.1
XM_005265031.2 Intron XP_005265088.2

View Full Product Details