Product Details

SNP ID
rs2054674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:123586690 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTTTTGAGCAACTTGTAAGAAACC[A/G]TTTTTAAGGCGGAAGTCGGGAATTT
Phenotype
MIM: 615939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HACD2 PubMed Links

Gene Details

Gene
HACD2
Gene Name
3-hydroxyacyl-CoA dehydratase 2
There are no transcripts associated with this gene.

Gene
MYLK-AS1
Gene Name
MYLK antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details