Product Details

SNP ID
rs10187018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203024135 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTTGAGCTCGGGAGTTTCAGTCT[A/G]CAGTGAGCTGGGATCATGCCACTCT
Phenotype
MIM: 609816
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NBEAL1 PubMed Links

Gene Details

Gene
NBEAL1
Gene Name
neurobeachin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114132.1 Intron NP_001107604.1
XM_005246787.3 Intron XP_005246844.1
XM_005246788.2 Intron XP_005246845.1
XM_006712698.3 Intron XP_006712761.1
XM_006712699.3 Intron XP_006712762.1
XM_011511658.2 Intron XP_011509960.1
XM_011511659.2 Intron XP_011509961.1
XM_011511660.2 Intron XP_011509962.1
XM_011511662.2 Intron XP_011509964.1
XM_011511663.2 Intron XP_011509965.1
XM_017004728.1 Intron XP_016860217.1
XM_017004729.1 Intron XP_016860218.1

View Full Product Details