Product Details

SNP ID
rs35370338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58729068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGCCCATATCCCAGCTCAAGT[A/G]AAACCTTCAGATGGCTGCAGTCTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf67 PubMed Links

Gene Details

Gene
C3orf67
Gene Name
chromosome 3 open reading frame 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198463.2 Intron NP_940865.1
XM_005264929.2 Intron XP_005264986.1
XM_006713013.3 Intron XP_006713076.1
XM_011533451.2 Intron XP_011531753.1
XM_011533453.2 Intron XP_011531755.1
XM_011533455.2 Intron XP_011531757.1
XM_011533456.1 Intron XP_011531758.1
XM_011533457.1 Intron XP_011531759.1
XM_011533458.2 Intron XP_011531760.1
XM_011533459.2 Intron XP_011531761.1
XM_011533460.2 Intron XP_011531762.1
XM_011533461.1 Intron XP_011531763.1
XM_011533463.2 Intron XP_011531765.1
XM_017005848.1 Intron XP_016861337.1
XM_017005849.1 Intron XP_016861338.1
XM_017005850.1 Intron XP_016861339.1
XM_017005851.1 Intron XP_016861340.1

View Full Product Details