Product Details

SNP ID
rs10935213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136974617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTATAATGACCATTTTACATTGT[A/G]TCTCTCTGGACATCATGAGCCTCCT
Phenotype
MIM: 605621
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL20RB PubMed Links

Gene Details

Gene
IL20RB
Gene Name
interleukin 20 receptor subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144717.3 Intron NP_653318.2
XM_006713665.3 Intron XP_006713728.1
XM_011512910.2 Intron XP_011511212.1
XM_011512911.2 Intron XP_011511213.1
XM_017006632.1 Intron XP_016862121.1

View Full Product Details