Product Details
- SNP ID
-
rs2077887
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:174057534 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTAGCTGGCTGGTTGTAGTGACTT[C/T]GTAGGGTTGTATTTTCCCTGCTCTC
- Phenotype
-
MIM: 616752
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HMP19
PubMed Links
Gene Details
- Gene
- HMP19
- Gene Name
- HMP19 protein
There are no transcripts associated with this gene.
View Full Product Details