Product Details
- SNP ID
-
rs7306771
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:104067156 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGTTGGCATTGGTATCAACTGACA[G/T]ATTCACAACTTTGAATAAAACTTTC
- Phenotype
-
MIM: 607926
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GLT8D2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112883159] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLT8D2
- Gene Name
- glycosyltransferase 8 domain containing 2
There are no transcripts associated with this gene.
- Gene
- HCFC2
- Gene Name
- host cell factor C2
View Full Product Details