Product Details

SNP ID
rs536662
Assay Type
Functionally tested
NCBI dbSNP Submissions
70
Location
Chr.1:119750110 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGAACCTCATTGCTTCTCCCCAGAA[A/G]GAATCCCATGATATAAGAATTGTTA
Phenotype
MIM: 600234 MIM: 606879
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMGCS2 PubMed Links

Gene Details

Gene
HMGCS2
Gene Name
3-hydroxy-3-methylglutaryl-CoA synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166107.1 Intron NP_001159579.1
NM_005518.3 Intron NP_005509.1
XM_011541313.1 Intron XP_011539615.1
Gene
PHGDH
Gene Name
phosphoglycerate dehydrogenase
There are no transcripts associated with this gene.

View Full Product Details