Product Details

SNP ID
rs11586225
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:38891303 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAGAAGAATATTTACAACTTTAT[A/C]CACATTACATATTACATATTACAAA
Phenotype
MIM: 611923 MIM: 608962
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GJA9 PubMed Links

Gene Details

Gene
GJA9
Gene Name
gap junction protein alpha 9
There are no transcripts associated with this gene.

Gene
GJA9-MYCBP
Gene Name
GJA9-MYCBP readthrough
There are no transcripts associated with this gene.

Gene
RHBDL2
Gene Name
rhomboid like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304746.1 Intron NP_001291675.1
NM_017821.4 Intron NP_060291.2

View Full Product Details