Product Details

SNP ID
rs9526736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51225361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGACTAAATCAACTGAAGGTAA[C/T]AGCAAGCACTTTCTTAGCATTTAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM124A PubMed Links

Gene Details

Gene
FAM124A
Gene Name
family with sequence similarity 124 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242312.1 Intron NP_001229241.1
NM_145019.3 Intron NP_659456.3
XM_011534978.2 Intron XP_011533280.1
XM_017020419.1 Intron XP_016875908.1
XM_017020420.1 Intron XP_016875909.1

View Full Product Details