Product Details

SNP ID
rs3317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:112876454 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGTATCACATCTTCAGGATAGGT[A/G]ATAACAGTGTGAAGGGTGTGCTCAT
Phenotype
MIM: 125265 MIM: 182175
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
REEP5 PubMed Links

Gene Details

Gene
REEP5
Gene Name
receptor accessory protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005669.4 3046 UTR 3 NP_005660.4
XM_017009844.1 3046 Intron XP_016865333.1
Gene
SRP19
Gene Name
signal recognition particle 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204193.1 3046 Intron NP_001191122.1
NM_001204194.1 3046 Intron NP_001191123.1
NM_001204196.1 3046 Intron NP_001191125.1
NM_001204199.1 3046 Intron NP_001191128.1
NM_003135.2 3046 Intron NP_003126.1

View Full Product Details