Product Details
- SNP ID
-
rs12516305
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:123091526 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACGCTTATCACAAGTACAGTTCCA[G/T]TTGGCGGTTTTGAAGCAAAACTCTA
- Phenotype
-
MIM: 616982
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRDM6
PubMed Links
Gene Details
- Gene
- PRDM6
- Gene Name
- PR domain 6
View Full Product Details