Product Details

SNP ID
rs11882783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50337927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCCCAATGGTATCCAAGTCCTT[A/G]AAATAAGCCATTCCCAGGGAATTGA
Phenotype
MIM: 176264
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNC3 PubMed Links

Gene Details

Gene
KCNC3
Gene Name
potassium voltage-gated channel subfamily C member 3
There are no transcripts associated with this gene.

Gene
NAPSB
Gene Name
napsin B aspartic peptidase, pseudogene
There are no transcripts associated with this gene.

View Full Product Details