Product Details

SNP ID
rs3734123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:136225445 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGGACTTGAACAGTATCCATATA[C/T]GGGGCTGTCTCAGTGAAGCAGCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPC7 PubMed Links

Gene Details

Gene
TRPC7
Gene Name
transient receptor potential cation channel subfamily C member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167576.1 Intron NP_001161048.1
NM_001167577.1 Intron NP_001161049.1
NM_020389.2 Intron NP_065122.1
Gene
TRPC7-AS2
Gene Name
TRPC7 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details